Thermo Scientific 9863BN pH microelectrode, Needle Tip, 3.3-ft (1-m)
Representative image only.
Thermo Scientific Mfr # 9863BN – Item # WZ-05719-24
Designed to measure samples as small as 0.5 uL in containers as small as 384 well plates
  • Ideal for use with smaller sample sizes and are typically combination electrodes
  • Use micro pH electrodes in conjunction with many instruments including capillary tubes and microwell plates
  • 1 year warranty
PRODUCT OPTIONS
£821.35GBP / Each
£985.62GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Electrode TypeSingle junction
  • Electrode Body MaterialGlass
  • Reference JunctionCeramic
  • Reference ElectrolyteAg/AgCl
  • Min pH (pH)14
  • Min Temperature (° C)0
  • Max Temperature (° C)80
  • ConnectorBNC
  • Diameter (mm)1.27
  • Length (mm)40
  • Cable Length (m)1
  • DescriptionpH microelectrode, Needle Tip, 3.3-ft (1-m)
  • Warranty1 year
1 Year Warranty
Key Features
  • Ideal for use with smaller sample sizes and are typically combination electrodes
  • Use micro pH electrodes in conjunction with many instruments including capillary tubes and microwell plates
  • 1 year warranty
More About this Item
Microtip fits into small vessels and requires low volumes to obtain pH measurements.