This item is now discontinued. Please browse similar items
Thermo Scientific 66330 Slide-A-Lyzer Dialysis Cassettes, 3500 MWCO, 0.5-3 mL; 10/Pk
Representative image only.
Thermo Scientific Mfr # 66330 – Item # WZ-02905-03
High Performance Dialysis require just half the time of dialysis tubing for sample size from 100µl to 30mL
  • The innovative cassette design does not require the use of knots or clips that can lead to leaking sample loss.
  • Color coded translucent frame permits smooth sample withdrawal of sub-filter volumes ¨C removing every last drop is easy.
  • High surface area/sample volume ratio will dialyze twice as fast as dialysis via conventional tubing.
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialRegenerated Cellulose
  • Max Molecular Weight Cut Off (Mwco) (Dalton)3500
  • Max Sample Volume (mL)3
  • Min Sample Volume (mL)0.5
  • ColorPink
  • DescriptionSlide-A-Lyzer Dialysis Cassettes, 3500 MWCO, 0.5-3 mL; 10/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • The innovative cassette design does not require the use of knots or clips that can lead to leaking sample loss.
  • Color coded translucent frame permits smooth sample withdrawal of sub-filter volumes ¨C removing every last drop is easy.
  • High surface area/sample volume ratio will dialyze twice as fast as dialysis via conventional tubing.
  • 1 year warranty
More About this Item
Slide-A-Lyzer® dialysis cassette is ideal for low molecular weight contaminant removal, buffer exchange, desalting and concentration. The cassette membrane is composed of low-binding regenerated cellulose and features a hermetically sealed sample chamber to maintain the highest possible sample retention. Sample introduction and removal are easily accomplished by penetrating the gasket with a hypodermic needle attached to a syringe. The gasket imparts resealability to the needle puncture, ensuring that no sample is lost from the cassette during dialysis.