Thermo Scientific 5954 Storage boxes for Freezers, 2"H; 12/Pk

Representative image only.
Thermo Scientific Mfr # 5954 – Item # WZ-44199-70
Safely store samples in your freezer
  • Constructed of durable fiberboard
  • Boxes measure 5" x 5" x 2" (12.7 x 12.7 x 5 cm)

£74.74GBP / Dozen of 1
£89.69GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Freezer Capacity (cu ft)13.4
  • Freezer Capacity (Liters)379
  • Max Temperature - Freezer (° C)-50
  • Power (VAC)208 / 230
  • Power (Hz)60
  • Power (Breaker)15 A
  • DescriptionStorage boxes for Freezers; 2\"H

Key Features

  • Constructed of durable fiberboard
  • Boxes measure 5" x 5" x 2" (12.7 x 12.7 x 5 cm)

More About this Item

Store a variety of vials and tubes with Thermo Scientific™ Storage Boxes, for use in ultra-low temperature freezers.

ACCESSORIES

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat