This item is now discontinued. Please browse similar items
Thermo Scientific 2065Q Microtiter Plate Heating Block
Representative image only.
Thermo Scientific Mfr # 2065Q – Item # WZ-03137-10
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Height (in)1 3/4
  • Width (in)3 3/4
  • Depth (in)5 7/8
  • DescriptionMicrotiter Plate Heating Block
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Blocks accommodate any 6- to 96-well plate (round or flat bottom). Aluminum construction provides excellent heat transfer; model 03137-08 provides direct contact between the block heater and the bottom of microtiter plates for greater temperature uniformity. Color changes can easily be viewed against the white anodized surfaces. Finger indentations allow easy removal. Overall dimensions are 3-3/4"W x 1-3/4"H x 5-7/8"D.