This item is now discontinued. Please browse similar items
Testo 0635 0545 Light Intensity Probe; 0 to 100,000 Lux
Representative image only.
Testo Mfr # 0635 0545 – Item # WZ-37486-09
Optimize performance of your air/gas analyzer
  • Replacement probes
  • Data logging software
  • White or warm light LEDs (K< 5,000) can be measured without any problem
  • Accuracy according to DIN EN 13032-1 and class C according to DIN 5032-7
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithMultifunction air measurement meter 37489-01
  • DescriptionLight Intensity Probe; 0 to 100,000 Lux
Two year warranty
Key Features
  • Replacement probes
  • Data logging software
  • White or warm light LEDs (K< 5,000) can be measured without any problem
  • Accuracy according to DIN EN 13032-1 and class C according to DIN 5032-7
  • 2 year warranty
More About this Item
Replacement probes, software, and other accessories to customize your sampling and analysis system.
Use the lux probe to check illuminance. It is suitable for all common lamps and light sources, because the assessment is carried out according to the V-lambda curve which corresponds to the sensitivity of the human eye in daylight. In addition, the illuminance and image refresh rate can be determined on CRT monitors. This probe is suitable for measuring light intensity in conjunction with the 37489-01 air measurment meter, meter sold separately. This probe has a measuring range of 0 to 100,000 lux (0 to 300 Hz) with a 1 lux resolution and has a 1.8 m fixed cable.