This item is now discontinued. Please browse similar items
Testo 0603 3392 Type T Thermocouple SS Food Probe, -58 to 662F, PTFE CableIP-67 Rated
Representative image only.
Testo Mfr # 0603 3392 – Item # WZ-90025-27
Accurate Type T thermocouple probes idea for food applications
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Meter Measurement TypeThermocouple
  • Types Of Probes AcceptedType T
  • Min Temperature (° F)-58
  • Max Temperature (° F)662
  • Min Temperature (° C)-50
  • Max Temperature (° C)350
  • DescriptionType T Thermocouple SS Food Probe, -58 to 662F, PTFE CableIP-67 Rated
  • Warranty2 year
Two year warranty
Key Features
  • 2 year warranty
More About this Item
The Testo 0628 0027 waterproof Type T thermocouple needle probe measures temperature from -50 to 250 degrees C (-58 to 482 degrees F). Probe measures 150mm (5.9") and has a 1.4mm (0.05") diameter tip for reduced impact and features a response time of 2 seconds for fast readings.
The Testo 0603 3392 IP67 waterfproof food probe measures temperature from -50 to 350 degrees C (-58 to 662 degrees F). Probe measures 4.9" long and has a 0.16" diameter. This probe features a response tome of 7 seconds.
The Testo 0603 2192 IP67 waterfproof stainless steel probe measures temperature from -50 to 350 degrees C (-58 to 662 degrees F). Probe measures 4.9" long and has a 0.16" diameter and comes with a PUR cable. This probe features a response tome of 7 seconds.