This item is now discontinued. Please browse similar items
Techne FWSP660I Temperature Probe with Case and Calibration Cert, PRT, 1/4" dia, 660C
Representative image only.
Techne Item # WZ-16115-91
Optimize performance and protection of your Techne baths
  • Calibrated Accuracy of ±0.010 at 0°C
  • Calibrated range of -196°C to 660°C
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTemperature Probe with Case and Calibration Cert, PRT, 1/4\" dia, 660C
  • Warranty1 Year
1 Year Warranty
Key Features
  • Calibrated Accuracy of ±0.010 at 0°C
  • Calibrated range of -196°C to 660°C
  • 1 year warranty
More About this Item
The Techne Secondary Standard Probes offer proven and reliable performance for your critical temperature measurements. It can be used with the Tecal Accu-Temp high accuracy digital thermometer or any comparable unit. This probe includes a hard carrying case and ITS-90 (NIST traceable) calibration certificate with coefficients.
Provides a higher level of accuracy and stability than the Techne WSP probes making it the ideal choice for use as a secondary standard reference thermometer.
Secondary standard 25 or 100 ohm SPRT diameter 1/4 inch x 18 inch long including handle.