This item is now discontinued. Please browse similar items
Techne FWSP350I Temperature Probe with Case and Calibration Cert, PRT, 1/4" dia, 350C
Representative image only.
Techne Item # WZ-16115-90
Optimize performance and protection of your Techne baths
  • 350°C working standard probe
  • 1/4" diameter by 12" long
  • Includes case and calibration certificate
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTemperature Probe with Case and Calibration Cert, PRT, 1/4\" dia, 350C
  • Warranty1 Year
1 Year Warranty
Key Features
  • 350°C working standard probe
  • 1/4" diameter by 12" long
  • Includes case and calibration certificate
  • 1 year warranty
More About this Item
The Techne Working Standard Probes are an ideal choice for use in verifying, calibrating and monitoring the temperature of calibration baths, furnaces, precision fluidised baths, Dri-block calibrators, incubators, freezers, water baths, thermal validation systems, comparison calibrations of other sensors or for any other temperature application where a high level of accuracy and reliability are required.
All probes shown are fully annealed to remove any instability for longer term accuracy and performance throughout the operating range. Working standard 100 ohm PRT diameter 1 / 4 inch x 12 inch long with hard carrying case including ITS90 calibration certificate with coefficients. Calibrated Accuracy ±0.025 at 0°C. Calibrated range -50 to 350°C or 425°C.