This item is now discontinued. We recommend the following as suggested replacements: WZ-93947-00
Techne Prime Pro 48 Real-Time PCR System; 100-240 V, 50/60 Hz
Representative image only.
Techne Item # WZ-93945-14
Get quicker, more precise cycles from one temperature to the next
  • Fast cycling — runs 40 cycles in under 20 minutes when optimized
  • Process up to 40% more samples per hour than a traditional 96-well instrument
  • Up to 5x greater uniformity than any other block-based qPCR instrument
  • Uses four colors for easy multiplexing
  • Market-leading uniformity ±0.1°C at 95°C
  • 2 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity48-well plate, 5 to 20 µL/well
  • Max Temperature (° C)100
  • Min Temperature (° C)30
  • Temperature Accuracy±0.25⁰C
  • Max Heating Rate5.5⁰C per second
  • Block Uniformity±0.1⁰C
  • MemoryUnit saves the run data of last experiment, or unlimited via USB
  • DescriptionReal-Time PCR System; 100-240 V, 50/60 Hz
  • Warranty2 year
Two year warrantyConformite EuropeenneCanadian Standards Association
Key Features
  • Fast cycling — runs 40 cycles in under 20 minutes when optimized
  • Process up to 40% more samples per hour than a traditional 96-well instrument
  • Up to 5x greater uniformity than any other block-based qPCR instrument
  • Uses four colors for easy multiplexing
  • Market-leading uniformity ±0.1°C at 95°C
  • 2 year warranty
More About this Item
Compact real-time thermal cycler can accommodate a 48-well polypropylene PCR plate utilizing the same geometry as standard 384-well plates, but is only 1/8 of the size. It can process up to 40% more samples per hour than a traditional 96-well instrument. Suitable for both single labs and core facilities, the thermal cycler’s fast cycling enables several experiments per day. A specially designed thermal block delivers a unique heating and cooling system that provides accurate ±0.1°C temperature control and quickly cycles from one temperature to the next. An advanced high-performance optical system delivers precise and sensitive fluorescence detection, facilitating all four color multiplex applications. The qPRC unit is MIQE compliant, features an adaptive LED control and high-resolution melt (HRM) functionality. Easy-to-use software provides instrument control, data collection, and data analysis. An ergonomic plate loading dock allows for quick loading of the plate and is backlit for clear visibility.
Prime Pro 48 also accepts all popular brands of reagents. Always reads all 48 wells, with all 4 filters at every cycle, and automatically normalizes fluorescence—Never saturate the detector. Eliminate cross talk.
The ProDock (included) makes cell loading easier. The ergonomic platform fits into standard centrifuge rotors and is backlit for sample visualization—also works well with 16-channel pipettes.