This item is now discontinued. Please browse similar items
Techne DNA Shiga toxin (stx1) producing Escherichia coli, with mastermix
Representative image only.
PCRmax Item # WZ-93954-87
Screen for systemic microbial pathogens quickly to prevent economic loss
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
SPECS & DESCRIPTION
Specifications & Description
  • Test TypeDNA
  • Number of Tests Included150
  • Mastermix IncludedYes
  • Genus DetectedShiga
  • Binomial NameShiga toxin (stx1) producing Escherichia coli
  • DescriptionShiga toxin (stx1) producing Escherichia coli, with mastermix
Includes
Lyophilized mastermix, lyophilized ROX, resuspension buffer, lyophilized primer and probe mix, positive control for standard curve, internal extraction control DNA, endogenous control, and RNAse/DNAse-free water.
Key Features
  • No need to refrigerate—all contents are lyophilized for simple transport and storage
  • Positive copy number standard curve provides accurate quantification
  • Exceptional value at 150 tests per kit
More About this Item
Prevent economic loss, especially in the dairy and agricultural industry, using these microbial detection kits to accurately assess whether these disease-causing pathogens are present. Select a complete kit with mastermix or order a kit without mastermix and use your own.