This item is now discontinued. Please browse similar items
Taylor 503 Connoisseur Series Oven / Grill Analog Dial Thermometer with Silicone Grips
Representative image only.
Taylor Mfr # 503 – Item # WZ-90001-46
Get more reliable, consistent results
  • Temperature ranges from 150 to 600°F.
  • Large 3-1/2" thermometer head with a 2-1/2" viewing dial for easy reading.
  • Stainless steel construction with silicone grips for safe and easy handling.
  • Three-position alignment allows the thermometer to hang, stand, or clip to an oven rack
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Min Temperature (° F)150
  • Max Temperature (° F)600
  • Max Temperature (° C)316
  • Division (° F)5
  • Dial Size (in)2.5
  • DescriptionOven / Grill Analog Dial Thermometer with Silicone Grips
  • Warranty1 year
1 Year Warranty
Key Features
  • Temperature ranges from 150 to 600°F.
  • Large 3-1/2" thermometer head with a 2-1/2" viewing dial for easy reading.
  • Stainless steel construction with silicone grips for safe and easy handling.
  • Three-position alignment allows the thermometer to hang, stand, or clip to an oven rack
  • 1 year warranty
More About this Item
Oven thermometer. Since most oven temperatures vary from top to bottom and side to side, place an oven thermometer next to food while cooking to ensure proper temperatures. Connoisseur Series Bimetal Oven Thermometer features a compact dial with guidelines for warming, baking, and roasting. The durable stainless steel and glass lens construction is enhanced by silicone accents to make handling easier with wet hands or gloves. Rotating base clips, hangs or stands. Leave in oven to monitor temperatures. Hand wash only.