This item is now discontinued. Please browse similar items
Taylor 44S369 Squeeze Bulb Pressure Calibrator, with Case, 0 to 160" H2O
Representative image only.
Taylor Mfr # 44S369 – Item # WZ-68454-74
Small size allows you to test the output pressure of a pneumatic instrument and make calibration adjustments at the same time
SPECS & DESCRIPTION
Specifications & Description
  • Max Pressure160 inch of water
  • Min Pressure0 in of water
  • DescriptionSqueeze Bulb Pressure Calibrator, with Case, 0 to 160\" H2O
More About this Item
This calibrator allows a technician to test the output or differential pressure (flow) readings of a pneumatic instrument with one hand allowing the other hand to make calibration adjustments on the instrument being tested. The calibrator weighs only 8 oz. as compared to 3 or more lbs. for equipment marketed in kit form or one made up by the technician from parts.
The calibrator consists of sphygmomanometer-type gauge accurate to 1% with a squeeze bulb for applying pressure manually and a precision-machined hand-adjusted valve to hold and relieve the calibrating pressure. A flexible hose and a 3/16" tube-1/4" NPT compression fitting is supplied for quick and easy connection to unit being tested.
CALIBRATES
  • Pneumatic controllers
  • Low pressure gauges
  • Pneumatic transmitters
  • Flow transmitters
  • REVIEWS