This item is now discontinued. We recommend the following as suggested replacements: WZ-62990-06
Tapered Cork Stoppers, Standard Size 1; 100/Pk
Representative image only.
Item # WZ-06300-05
Standard tapered stoppers, manufactured with XXXX-grade cork
SPECS & DESCRIPTION
Specifications & Description
  • Standard Size Number1
  • Top Outside Diameter (OD) (mm)11
  • Bottom Outside Diameter (OD) (mm)8
  • Height (mm)16
  • MaterialCork
  • DescriptionTapered Cork Stoppers, Standard Size 1; 100/Pk
More About this Item
Economical, non-reactive stoppers are ideal for contact with nonaggressive or low concentration liquids or powders under average laboratory usage.