This item is now discontinued. Please browse similar items
Sussman SSB38002A High Pressure Line Water Feed for Stainless Steel Steam Generator
Representative image only.
Sussman Mfr # SSB38002A – Item # WZ-07601-62
Minimize contaminants—all wetted metal parts are stainless steel
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionHigh Pressure Line Water Feed for SS Steam Generator
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
This water feed system is used when the boiler operating pressure is equal to or greater than the incoming water line pressure. This system is compatible with Sussman boiler model SSB 12-60 (sold separately).