Supco 6" Circular Chart Recorder Paper, 7 Day, 40 to 110°F; 60/Pack
Representative image only.
Supco Mfr # E40110F7 – Item # WZ-80055-38
For Supco brand chart recorders
  • Circular and strip charts
  • For CRTH2 and CR4-KTH recorders
  • 1 year warranty
PRODUCT OPTIONS
£51.64GBP / Pkg of 60
£61.97GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Chart TypeCircular
  • Chart Speed7 day
  • Chart Diameter (in)6
  • Max Temperature (° F)110
  • Min Temperature (° F)40
  • Max Temperature (° C)43
  • Min Temperature (° C)4
  • Description6\" Chart Paper, 40 to 110 F, 7 Day; 60/Pk
  • Warranty1 year
1 Year Warranty
Key Features
  • Circular and strip charts
  • For CRTH2 and CR4-KTH recorders
  • 1 year warranty
More About this Item
Choose the paper compatible with your Supco chart recorder and application needs.