Cole-Parmer Vortex Mixer/Shaker Accessory Cup Pack
Representative image only.
Cole-Parmer Item # WZ-04729-07
Improve and expand the performance of you vortex mixer
  • 3 year warranty
£77.77GBP / Each
£93.32GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionVortex Mixer/Shaker Accessory Cup Pack
  • Warranty3 Years
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
For use with the variable-speed vortexer, pack contains:
1 x plastic cradle for standard microtitre plate
1 x circular foam insert for beakers and flasks (up to 500ml capacity)
1 x rectangular foam insert (holds 8 x 0.2ml, 8 x 0.5ml and 16 x 1.5ml microcentrifuge tubes)
1 x solid rectangular foam insert (blank for custom holes)
Please note: The microtitre plate holder SA8/1 should not be used at speeds above 800rpm