Cole-Parmer Tube Rotator Dual Tube Holder Disc Adapter
Representative image only.
Cole-Parmer Item # WZ-00077-23
Quickly and easily replace tube holder without tools
  • Choose the tube holder(s) that accommodates your tube size(s)
  • Fits two tube holders to Cole-Parmer® TR-200 series tube rotator simultaneously
  • 1 year warranty
  • Previous part number SB3/DUAL
PRODUCT OPTIONS
£95.44GBP / Each
£114.53GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionTube Rotator Dual Tube Holder Disc Adapter
  • Warranty1 Year
1 Year Warranty
Key Features
  • Choose the tube holder(s) that accommodates your tube size(s)
  • Fits two tube holders to Cole-Parmer® TR-200 series tube rotator simultaneously
  • 1 year warranty
  • Previous part number SB3/DUAL