Cole-Parmer Three Block or Dual Control Heater Cover
Representative image only.
Cole-Parmer Item # WZ-36610-97
Optimize performance of your Stuart block heaters
  • 1 year warranty
  • Previous part number SBH/3
PRODUCT OPTIONS
£147.81GBP / Each
£177.37GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionThree Block or Dual Control Heater Cover
  • Warranty1 year
1 Year Warranty
Key Features
  • 1 year warranty
  • Previous part number SBH/3
More About this Item
The heater covers ensure temperatures are maintained in your dry block heaters. Model 36610-96 cover is suitable for use with two block heaters. Model 36610-97 is suitable for use with three block or dual control heaters.
The extraction tool, 36610-98, makes removing blocks from the heater quick and effortless.