Cole-Parmer Microcentrifuge Tube Adaptors, 0.2 mL; 12/PK
Representative image only.
Cole-Parmer Item # WZ-85230-04
Enhance the performance of your Cole-Parmer® CF-200 Microcentrifuge
  • 3 year warranty
  • Previous part number SCF3/1/02
PRODUCT OPTIONS
£111.55GBP / Each
£133.86GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Tube Sizes Accepted0.2 mL
  • Number Of Tubes Accepted12
  • Adapter Shape TypeConical
  • DescriptionTube Adaptors, 0.2 mL; 12/pk
  • Warranty3 year
3 Year Warranty
Key Features
  • 3 year warranty
  • Previous part number SCF3/1/02
More About this Item
The Cole-Parmer CF-200 microcentrifuge includes a fixed-angle rotor that accommodates up to twelve 1.5- or 2.0-mL tubes. Order tube adapters to used 0.2- or 0.5-mL tubes, or order the strip tube rotor to accommodate two 8-place 0.2-mL strip tubes.