This item is now discontinued. Please browse similar items
Stuart Evaporator/Concentrator for Block Heaters
Representative image only.
Stuart Item # WZ-36610-99
Sample concentrator helps produce fast, efficient evaporation
  • 1 year warranty
Accessories for your block heater evaporator/concentrator
Accessories for Cole-Parmer® SC-200 Series Sample Concentrator
£19.05 - £856.88GBP / Each
£22.86 - £1,028.26GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionEvaporator/Concentrator for Block Heaters
  • Warranty1 Year
1 Year Warranty
Key Features
  • 1 year warranty
More About this Item
Use sample concentrator with three-block heaters 36610-12, -13, -22, and -23 to quickly concentrate multiple samples at once. Concentrator blows gas over the surface of your samples through stainless steel needles (not included). The heat from the block heater in combination with the blown gas produces ideal conditions for fast, efficient evaporation. Insert needles into a silicone membrane in virtually any configuration. Fully adjustable gas reservoir provides accurate height control. Order PTFE-coated needles for use with corrosive samples.