Cole-Parmer 8500 Tube Rotor, 1.5/2-mL; 8-Place
Representative image only.
Cole-Parmer Item # WZ-17414-71
Keep your Stuart centrifuge running smoothly with these replacement parts
  • Make the most of your centrifuge with interchangeable rotors and inserts
  • Designed to hold up to 8 microcentrifuge tubes with a 1.5 or 2 mL capacity
Call For Price
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)8500
  • Max RCF x g3884
  • Rotor TypeFixed Angle
  • Tube Sizes Accepted1.5 to 2 mL
  • Number Of Tubes Accepted8
  • DescriptionTube Rotor, 1.5/2-mL; 8-Place
Key Features
  • Make the most of your centrifuge with interchangeable rotors and inserts
  • Designed to hold up to 8 microcentrifuge tubes with a 1.5 or 2 mL capacity
More About this Item
These replacement parts are compatible with the Stuart® 8500 microcentrifuge, model 1741-70. Order microcentrifuge separately.