This item is now discontinued. Please browse similar items
Cole-Parmer 8500 Strip Tube Rotor, 0.2-mL PCR; 4 x 8-Place
Representative image only.
Cole-Parmer Item # WZ-17414-72
Keep your Stuart centrifuge running smoothly with these replacement parts
  • Make the most of your centrifuge with interchangeable rotors and inserts
  • Designed with 4 x 8-place descign to hold 0.2 mL PCR tube strips
SPECS & DESCRIPTION
Specifications & Description
  • Max Speed (rpm)8500
  • Max RCF x g3884
  • Rotor TypeStrip Tube
  • Tube Sizes Accepted0.2 mL PCR
  • Number Of Tubes Accepted32
  • DescriptionStrip Tube Rotor, 0.2-mL PCR; 4 x 8-Place
Key Features
  • Make the most of your centrifuge with interchangeable rotors and inserts
  • Designed with 4 x 8-place descign to hold 0.2 mL PCR tube strips
More About this Item
These replacement parts are compatible with the Stuart® 8500 microcentrifuge, model 1741-70. Order microcentrifuge separately.