Cole-Parmer VapLock™ Solvent Waste Manifold, PP, Justrite PP, 1x1/2" NPTF, 3x1/4" NPTF; 1/ea
Representative image only.
Cole-Parmer Item # WZ-12019-56
Connect solvent waste manifold caps to waste containers for a secure closed system
  • Choose from polypropylene or PTFE construction
PRODUCT OPTIONS
£195.41GBP / Each
£234.49GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolypropylene(PP)
  • Cap / Stopper SizeJustrite Quick-Disconnect, PP
  • Special Features1x1/2\" NPTF, 3x1/4\" NPTF
  • DescriptionSolvent Waste Manifold, PP, Justrite PP, 1x1/2\" NPTF, 3x1/4\" NPTF; 1/ea
Key Features
  • Choose from polypropylene or PTFE construction
More About this Item
Select from various manifold cap sizes to fit your laboratory and industrial solvent waste containers. Quick disconnect caps are available with PP or SS fittings and allow fast and easy connections to Justrite® safety containers. Caps are available with polypropylene (PP) or PTFE manifolds and 4 or 10 variously sized ports. Ports accept male thread adapters to connect flexible and rigid tubing, or filter 12019-79 or filter/indicator 12019-80.