Cole-Parmer VapLock™ Solvent Delivery Cap, four 1/4"-28 ports, GL32; 1/ea
Representative image only.
Cole-Parmer Item # WZ-12018-53
Reduce chemical evaporation and exposure to hazardous vapors
  • Use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) OD
PRODUCT OPTIONS
£46.41GBP / Each
£55.69GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Cap MaterialPTFE/PP
  • Cap / Stopper SizeGL32
  • Cap ColorBlue/white
  • Special Features4 x 1/4-28 UNF(F)
  • AutoclavableYes
  • DescriptionSolvent Delivery Cap, four 1/4-28 ports, GL32
Key Features
  • Use with semi-rigid tubing equal to or less than 1/8" (3.2 mm) OD
More About this Item
Create a closed system using solvent reservoir caps to prevent hazardous vapors from escaping into the lab. Caps accept standard 1/4"-28 UNF(M) fittings to easily connect semi-rigid tubing. Anti-twist design allows the inner body of the cap to rotate independently preventing tubes from tangling when caps are removed or attached. Caps are constructed of PTFE body and PP collar* and are autoclavable.
Caps with threaded port have ports angled at 10° tilt for more accessible tubing connections. Select caps with stainless steel (SS) thread insert to minimize cross-threading and damage to soft PTFE ports.
*Does not come in contact with liquid under normal circumstances.