Cole-Parmer VapLock™ Port Plug, Black Acetal, 1/4"-28 UNF(M); 10/PK
Representative image only.
Cole-Parmer Item # WZ-12020-46
Use plugs to seal unused ports for a leak-resistant system
  • Seal unused manifold and bottle cap ports
PRODUCT OPTIONS
£22.41GBP / Pkg of 10
£26.89GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Fitting MaterialPOM
  • Connection TypeUNF(M)
  • Thread size UNF(M) (port 1) (in)1/4\"-28
  • ColorBlack
  • DescriptionPort Plug, 1/4\"-28 UNF(M), black acetal; 10/pk
Key Features
  • Seal unused manifold and bottle cap ports
More About this Item
Bottom-sealing UNF thread options include 1/4-28 and 5/16-24, and do not require PTFE tape for leak-resistant connections.
Polyoxymethylene (POM), also known as acetal, is a strong plastic, ideal for rugged applications.