Spex VapLock™ Mixing Valve, 6-Position, 0.032" Orifice, PTFE Body and Diaphragm, 24 VDC
Representative image only.
Spex Mfr # 080T624-32 – Item # WZ-12021-59
Chemical-resistant materials allow for mixing of harsh fluids
  • High efficiency solenoid
  • Valve life above 2 million cycles
  • Chemical-resistant PTFE or PEEK body with PTFE diaphragm
  • Low power requirement and minimal heat generation
  • Maximum operating pressure of 20 psi
PRODUCT OPTIONS
£989.91GBP / Each
£1,187.89GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Outlet Port53 µL
  • Connection TypePTFE coated lead wires
  • Inlet Port15 µL
  • Housing MaterialPTFE
  • Electrical Connections24 VDC
  • Flow Pattern6 positions
  • DescriptionMixing Valve, 6-Position, 0.032\" Orifice, PTFE Body and Diaphragm, 24 VDC
Key Features
  • High efficiency solenoid
  • Valve life above 2 million cycles
  • Chemical-resistant PTFE or PEEK body with PTFE diaphragm
  • Low power requirement and minimal heat generation
  • Maximum operating pressure of 20 psi
More About this Item
These normally closed solenoid mixing valves feature low power consumption, minimal dead volume, and fast response time for all your mixing needs. All valves come with 15" (26 gauge) PTFE-coated lead wires.
When used as flow mixing devices, the flow from the inlets can be mixed together into a single stream under many different conditions. Each inlet can be selected individually. Any combination of inlets can be selected which results in the outlet being a mixture of the chosen inlets. Outlet composition will depend on pressure and fluid viscosity in each stream.
When the flow pattern is reversed, you can divert flow from a common port to one or multiple outlets. In flow selection applications, the common inlet is diverted to one of the available outlets. For stream splitting, the common inlet is split between the available outlets in a fixed or adjustable percentage. The split can be controlled from one outlet up to the maximum number of outlets in that valve.