Cole-Parmer Spex® 2231-10 Sealing Mat for Square 24-Well Microlate; 10/PK
Representative image only.
Cole-Parmer Item # WZ-04576-95
Tightly seals microplate wells to prevents spills and well-to-well contamination
  • Constructed of rugged silicone rubber
  • May be sterilized and reused
  • Use with 24-well microplates with square wells 04575-68 and 04576-94
  • Pack of 10
PRODUCT OPTIONS
£103.97GBP / Case of 10
£124.76GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSilicone
  • Compatible WithMicroplates 04575-68 and 04576-94
  • DescriptionSealing Mat for Square 24-Well Microlate; 10/PK
Key Features
  • Constructed of rugged silicone rubber
  • May be sterilized and reused
  • Use with 24-well microplates with square wells 04575-68 and 04576-94
  • Pack of 10
More About this Item
Use these flexible deep-well sealing mats to reduce maximum well volume. 