Cole-Parmer Spex® 2221 Sealing Mat for Rectangular 48-Well Microplate; 1/EA
Representative image only.
Cole-Parmer Item # WZ-04575-67
Tightly seals microplate wells to prevents spills and well-to-well contamination
  • Constructed of rugged silicone rubber
  • May be sterilized and reused
  • Use with 48-well microplates with rectangular wells (04575-66 and 04576-92)
  • 1 each
PRODUCT OPTIONS
£10.79GBP / Each
£12.95GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialSilicone
  • Compatible WithMicroplates 04575-66 and 04576-92
  • DescriptionSealing Mat for Rectangular 48-Well Microplate; 1/EA
Key Features
  • Constructed of rugged silicone rubber
  • May be sterilized and reused
  • Use with 48-well microplates with rectangular wells (04575-66 and 04576-92)
  • 1 each
More About this Item
Use these flexible deep-well sealing mats to reduce maximum well volume. 