Cole-Parmer Spex® 6885E Replacement End plugs for Mid-Size Grinding Vial Set 61043-13, PC; 2/PK
Representative image only.
Cole-Parmer Item # WZ-61034-65
Keep spare items to ensure there is no down time
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Polycarbonate end plugs
  • Eliminate metal contamination
  • Ideal for grinding soft plant and animal tissues
  • Pack of 2
PRODUCT OPTIONS
£254.27GBP / Pkg of 2
£305.12GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolycarbonate
  • Compatible WithMid-size grinding vial set 61043-13
  • DescriptionReplacement End plugs for Mid-Size Grinding Vial Set 61043-13, PC; 2/PK
Key Features
  • Suitable for use with the Cole-Parmer® Freezer/Mill® grinding vial sets
  • Polycarbonate end plugs
  • Eliminate metal contamination
  • Ideal for grinding soft plant and animal tissues
  • Pack of 2
More About this Item
Optimize performance of your Cole-Parmer® Freezer/Mill® grinding vial sets with a wide range of grinding ball options suitable for your application.