Cole-Parmer Spex® 2240-PEF Preloaded Vial Set, Polyethylene, 5 mL, 24 Vials/Set; 10 Sets/CS
Representative image only.
Cole-Parmer Item # WZ-04576-97
Preloaded with steel grinding balls for fast and convenient sample preparation
  • Optimize performance of your HG-400 MiniG® and Geno/Grinder® homogenizers
  • Each vial is loaded with one 3/8" (9.5 mm) steel grinding ball 04500-21
  • 1.5 mL working volume per vial
PRODUCT OPTIONS
£421.37GBP / Case of 10
£505.64GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)5
  • Tube MaterialPolyethylene
  • Special FeaturesIncludes grinding balls
  • DescriptionPreloaded Vial Set, Polyethylene, 5 mL, 24 Vials/Set; 10 Sets/CS
Includes
240 vials with caps, 240 steel balls, 24 foam vial holders, and 24 polyethylene carriers.
Key Features
  • Optimize performance of your HG-400 MiniG® and Geno/Grinder® homogenizers
  • Each vial is loaded with one 3/8" (9.5 mm) steel grinding ball 04500-21
  • 1.5 mL working volume per vial
More About this Item
The 4, 5, and 15 mL vial sets come in a foam vial holder within a sturdy polyethylene carrier that has the same footprint and height as a deep-well titer plate. Carrier are used in pairs and fits adjustable clamp assembly 04605-08. Run up to four vial sets at a time for high-throughput tissue homogenization and cell lysis. Use frosted PE vials 04576-97 with Cryo-Block 04500-25 to preserve temperature sensitive samples during grinding. Polyethylene is compatible with commonly used reagents such as TRIzol® solution.
Run up to four vial sets at a time for high-throughput tissue homogenization and cell lysis. Vials can be used with Cryo-Block 04500-26 to preserve temperature sensitive samples during grinding.
Each vial set comes in a foam vial holder within a sturdy polyethylene carrier that has the same footprint and height as a deep-well microplate. Carrier are used in pairs and fits adjustable clamp assembly 04605-08. Use frosted PE vials with Cryo-Block 04500-25 to preserve temperature sensitive samples during grinding. Polyethylene is compatible with commonly used reagents such as TRIzol® solution.