Cole-Parmer Spex® 2301-100MB Prefilled Tubes with Screw Cap, 2 mL, 100 µm Molecular Biology Grade Silica Beads; 100/PK
Representative image only.
Cole-Parmer Item # WZ-04577-02
Tubes are prefilled with acid washed grinding beads for your convenience
  • Self standing tubes with screw-on cap
  • Prefilled with RNase/DNase-free treated 100 µm silica grinding beads to inactivate contaminating enzymes and remove fine particles and contaminants
  • Ideal for disrupting bacteria
  • Self-standing tubes
PRODUCT OPTIONS
£211.79GBP / Pkg of 100
£254.15GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Volume (mL)2
  • Tube MaterialPolyethylene
  • Cap TypeScrew on
  • Cap colorRed
  • Fill100 µm molecular biology grade silica beads
  • DescriptionPrefilled Tubes with Screw Cap, 2 mL, 100 µm Molecular Biology Grade Silica Beads; 100/PK
Includes
100 µm molecular biology grade silica beads.
Key Features
  • Self standing tubes with screw-on cap
  • Prefilled with RNase/DNase-free treated 100 µm silica grinding beads to inactivate contaminating enzymes and remove fine particles and contaminants
  • Ideal for disrupting bacteria
  • Self-standing tubes
More About this Item
Use prefilled tubes to break up samples to release the DNA, RNA, and proteins contained within cells. Select from zirconia or silica beads. Beads are acid washed and processed to provide optimum results.