Cole-Parmer Spex® 3623C Polished Tungsten Carbide Pellets, 31 mm; 2/Pk
Representative image only.
Cole-Parmer Item # WZ-04577-43
Optimize performance of your Cole-Parmer® hydraulic laboratory pellet press
  • Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more
  • For use with evacuable pellet die set 04576-20
  • Constructed of durable polished tungsten carbide
PRODUCT OPTIONS
£586.53GBP / Pkg of 2
£703.84GBP (incl VAT)
OFTEN BOUGHT WITH…
SPECS & DESCRIPTION
Specifications & Description
  • MaterialPolished Tungsten Carbide
  • Compatible With04577-46 and 04577-47 press; 04576-20 die set
  • Diameter (mm)31
  • DescriptionPolished Tungsten Carbide Pellets, 31 mm; 2/Pk
Key Features
  • Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more
  • For use with evacuable pellet die set 04576-20
  • Constructed of durable polished tungsten carbide
More About this Item
Ensure top results from your Cole-Parmer® hydraulic laboratory pellet press for pressing of sample pellets for XRF and other analytical techniques with these accessories and components. The accessories and replacement parts include the 13 mm, 31 mm, 35 mm, and 40 mm pellet die sets, different material pellets, sleeve-and plunger sets, and more.