Cole-Parmer Spex® 1690 MiniG® Replacement Adjustable Clamp Assembly for HG-400 Homogenizer
Representative image only.
Cole-Parmer Item # WZ-04575-40
Accommodates a full range of sample vials with an adjustable clamp
  • Optimize performance of your HG-400 MiniG® homogenizer
  • Accommodates up to 48 x 2 mL trays of vials when stacked
PRODUCT OPTIONS
£831.54GBP / Each
£997.85GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • MaterialMetal
  • DescriptionMiniG® Replacement Adjustable Clamp Assembly for HG-400 Homogenizer
Key Features
  • Optimize performance of your HG-400 MiniG® homogenizer
  • Accommodates up to 48 x 2 mL trays of vials when stacked
More About this Item
Multiple inserts are available for a variety of homogenizing applications.
Clamp assembly easily adjusts to fit various sizes of titer plates and vials.