Cole-Parmer Spex® 1668 MiniG® Homogenizer Cryo-Block, Aluminum, 6 x 15 mL PC Vials 04576-99; 1/EA
Representative image only.
Cole-Parmer Item # WZ-04575-37
Accommodates a full range of sample vials with an adjustable clamp
  • Optimize performance of your HG-400 MiniG® homogenizer
  • Aluminumvial holder helps maintain cold temperature during processing
  • Holds six 15 mL polycarbonate vials
  • Hollow base makes it lightweight
PRODUCT OPTIONS
£140.77GBP / Each
£168.92GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Use with GeneratorsMiniG® Homogenizer
  • MaterialAluminum
  • DescriptionMiniG® Homogenizer Cryo-Block, Aluminum, 6 x 15 mL PC Vials 04576-99; 1/EA
Key Features
  • Optimize performance of your HG-400 MiniG® homogenizer
  • Aluminumvial holder helps maintain cold temperature during processing
  • Holds six 15 mL polycarbonate vials
  • Hollow base makes it lightweight
More About this Item
Multiple inserts are available for a variety of homogenizing applications.
Compatible with polycarbonate vials 04576-99.