Cole-Parmer Spex® 2110M-96 Magnetic Tips; 96/PK
Representative image only.
Cole-Parmer Item # WZ-04576-86
Get the most out of your Cole-Parmer® homogenizer
  • Choose from a wide range of accessories for your homogenizer
  • Strong magnets easily remove grinding balls from microplates
  • Compatible with 5/32" (4 mm) balls 04500-19
  • Pack of 96
PRODUCT OPTIONS
£900.13GBP / Pkg of 96
£1,080.16GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Use with Generators5/32\" (4 mm) balls 04500-19
  • DescriptionMagnetic Tips; 96/PK
Key Features
  • Choose from a wide range of accessories for your homogenizer
  • Strong magnets easily remove grinding balls from microplates
  • Compatible with 5/32" (4 mm) balls 04500-19
  • Pack of 96
More About this Item
Optimize your homogenizer with these quality accessories.
Converts micropipette dispensers into pin magnets for removing grinding balls from microplates.