Cole-Parmer Spex® 2198 Homogenizer Foam Holder, 6 x 75 mL PC Jars 04575-72; 1/EA
Representative image only.
Cole-Parmer Item # WZ-04575-63
Securely hold vials in place while processing samples
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Ideal for grinding large samples
  • Process 6 samples at one time
PRODUCT OPTIONS
£52.66GBP / Each
£63.19GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Max Tube Size (mL)75
  • Tubes Held6
  • MaterialFoam
  • DescriptionHomogenizer Foam Holder, 6 x 75 mL PC Jars 04575-72; 1/EA
Key Features
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Ideal for grinding large samples
  • Process 6 samples at one time
More About this Item
Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.