Cole-Parmer Spex® 2197 Homogenizer Foam Holder, 24 x 15 mL Centrifuge Tubes; 1/EA
Representative image only.
Cole-Parmer Item # WZ-04575-62
Securely hold vials in place while processing samples
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Process 24 samples at one time
PRODUCT OPTIONS
£52.91GBP / Each
£63.49GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Max Tube Size (mL)15
  • Tubes Held24
  • MaterialFoam
  • DescriptionHomogenizer Foam Holder, 24 x 15 mL Centrifuge Tubes; 1/EA
Key Features
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Process 24 samples at one time
More About this Item
Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.