Cole-Parmer Spex® 2198-24-PE Homogenizer Foam Holder, 24 x 12 mL Vials; 1/EA

Representative image only.
Cole-Parmer Item # WZ-04576-89
Securely hold vials in place while processing samples
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Holds twenty-four 12 mL vials

PRODUCT OPTIONS

£37.52GBP / Each
£45.02GBP (incl VAT)

SPECS & DESCRIPTION

Specifications & Description

  • Max Tube Size (mL)12
  • Tubes Held24
  • MaterialFoam
  • DescriptionHomogenizer Foam Holder, 24 x 12 mL Vials; 1/EA

Key Features

  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Holds twenty-four 12 mL vials

More About this Item

Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.

REVIEWS

Q&A

CUSTOMER SUPPORT

Need Help with this product?

Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.

Call Us At+44 (0)1480 272279
Live chat