Cole-Parmer Spex® 2193 Homogenizer Foam Holder, 12 x 15mL Short PC Vials 04576-99; 1/EA
Representative image only.
Cole-Parmer Item # WZ-04575-61
Securely hold vials in place while processing samples
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Holds twelve 15 mL short vials 04576-99
PRODUCT OPTIONS
£58.01GBP / Each
£69.61GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Max Tube Size (mL)15
  • Tubes Held12
  • MaterialFoam
  • DescriptionHomogenizer Foam Holder, 12 x 15 mL Short PC Vials 04576-99; 1/EA
Key Features
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Holds twelve 15 mL short vials 04576-99
More About this Item
Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.