Cole-Parmer Spex® 2260 Homogenizer Cryo-Block, Aluminum, 8 x Grinding Vials 04575-73; 2/PK
Representative image only.
Cole-Parmer Item # WZ-04575-78
Securely hold vials in place while processing samples
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Process 16 samples simultaneously
  • Accepts grinding vials 04575-73
  • Enables high-throughput cryogenic grinding
  • Increases yields by grinding samples at sub-zero temperatures
PRODUCT OPTIONS
£906.02GBP / Pkg of 2
£1,087.22GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Tubes Held8
  • MaterialAluminum
  • DescriptionHomogenizer Cryo-Block, Aluminum, 8 x Grinding Vials 04575-73; 2/PK
Key Features
  • Safely grind samples in the HG-600 Geno/Grinder® 2010
  • Process 16 samples simultaneously
  • Accepts grinding vials 04575-73
  • Enables high-throughput cryogenic grinding
  • Increases yields by grinding samples at sub-zero temperatures
More About this Item
Vial holders are suitable for use with the HG-600 Geno/Grinder® 2010 Homogenizer. Order homogenizer and vials separately.
Each aluminum cryo-block holds eight grinding vials 04575-73. Chill cryo-blocks in the cryo-station 04575-81 to maintains samples at liquid nitrogen temperatures during tissue homogenization and cell lysis. The blocks preserve pesticides, RNA, and proteins for heat sensitive extraction methods. Use two at a time in the clamp assembly 04605-08 to keep it in balance. Geno/Grinder speed restrictions apply for running more than 8 vials simultaneously.