This item is now discontinued. Please browse similar items
Cole-Parmer Spex® 2051 Genomax Adjustable Clamp for Vials, Tubes, and Jars
Representative image only.
Cole-Parmer Item # WZ-41056-99
Quick and efficient extraction of nucleic acids, proteins, and other molecules through an innovative bead beating technique
  • Optimize performance of your GenoLyte® homogenizer and BM-200 Mixer/Mill® grinder
  • Accomodates up to 96 x 2 mL or 5 mL vials
  • Quick-locking system allows for easy installation and removal
SPECS & DESCRIPTION
Specifications & Description
  • Compatible WithTemperature controlled GenoLyte
  • DescriptionGenomax Adjustable Clamp for Vials, Tubes, and Jars
Key Features
  • Optimize performance of your GenoLyte® homogenizer and BM-200 Mixer/Mill® grinder
  • Accomodates up to 96 x 2 mL or 5 mL vials
  • Quick-locking system allows for easy installation and removal
More About this Item
Get the accessories you need to properly operate your HG-200 GenoLyte®.
The 2051 clamp adjusts to fit various sizes of vials. It holds up to 48 x 2 mL vials as a single layer, or 96 x 2 mL vials when stacked. The quick-locking system on the Genomax allows for easy installation and removal of the clamp. Users can load the clamp on the bench top and then lock it into place on the Genomax or leave the clamp in place and load the vials directly into the Genomax.