Cole-Parmer Spex® 1217 GenoLyte® Homogenizer Vial Holder, 1 x 3.5 mL Agate Vial 04575-93; 1/EA
Representative image only.
Cole-Parmer Item # WZ-04575-34
Quick and efficient extraction of nucleic acids, proteins, and other molecules through an innovative bead beating technique
  • Optimize performance of your GenoLyte® homogenizer and BM-200 Mixer/Mill® grinder
  • Holds one 3.5 mL agate vial 04575-93
PRODUCT OPTIONS
£729.33GBP / Each
£875.20GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Use with GeneratorsHG-200 GenoLyte® homogenizer
  • DescriptionGenoLyte® Homogenizer Vial Holder, 1 x 3.5 mL Agate Vial 04575-93; 1/EA
Key Features
  • Optimize performance of your GenoLyte® homogenizer and BM-200 Mixer/Mill® grinder
  • Holds one 3.5 mL agate vial 04575-93
More About this Item
Get the accessories you need to properly operate your HG-200 GenoLyte®.