Cole-Parmer Spex® 3616 Evacuable Pellet Die Set for 35 mm Sample Disks; 1/Ea
Representative image only.
Cole-Parmer Item # WZ-04576-14
Optimize performance of your Cole-Parmer® hydraulic laboratory pellet press
  • Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more
  • Used to form 35 mm sample disks
PRODUCT OPTIONS
£2,816.46GBP / Each
£3,379.75GBP (incl VAT)
OFTEN BOUGHT WITH…
SPECS & DESCRIPTION
Specifications & Description
  • Compatible With04577-46 and 04577-47 press
  • Diameter (mm)35
  • DescriptionEvacuable Pellet Die Set for 35 mm Sample Disks; 1/Ea
Includes
evacuable die body, base, plunger, two polished steel pellets, O-ring vacuum seals, and knock-out ring for sample disk extraction.
Key Features
  • Choose from a wide range of accessories including pellet die sets, pellets, flaring tool, and more
  • Used to form 35 mm sample disks
More About this Item
Ensure top results from your Cole-Parmer® hydraulic laboratory pellet press for pressing of sample pellets for XRF and other analytical techniques with these accessories and components. The accessories and replacement parts include the 13 mm, 31 mm, 35 mm, and 40 mm pellet die sets, different material pellets, sleeve-and plunger sets, and more.