Cole-Parmer Spex® 2205-50 Deep-Well Microplate, 96 x 1.4 mL, Square Wells, Reinforced; 50/CS
Representative image only.
Cole-Parmer Item # WZ-41010-96
Durable polypropylene microplates resist perforation by grinding media
  • Alphanumeric marks for well identification
  • Use microplates with sealing mats to prevent spills and well-to-well contamination
  • DNAse/RNAse free
  • Toughened
  • Round bottom
PRODUCT OPTIONS
£209.06GBP / Case of 50
£250.87GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Number of Wells96
  • Well Volume (mL)1.4
  • Max Working Well Volume (mL)1
  • Well ShapeSquare
  • Description96 Square Well Reinforced Titer Plate; 50/CS
Key Features
  • Alphanumeric marks for well identification
  • Use microplates with sealing mats to prevent spills and well-to-well contamination
  • DNAse/RNAse free
  • Toughened
  • Round bottom
More About this Item
Ideal for use with Cole-Parmer® homogenizers, these deep-well plates serve many application purposes, such as tissue homogenization and cell lysis.
Deep-well reinforced titer plate with a 96-well format. Each square well has a volume of 1.4 mL and a working volume of 1 mL. Compatible with the Geno/Grinder® due to the toughened wells that withstand bead beating. This plate can also be used with the HG-400 MiniG®.
Use with matching cap-mat 41010-97.