Cole-Parmer Spex® 2189C Cryoplate; 1/EA
Representative image only.
Cole-Parmer Item # WZ-04576-88
Get the most out of your Cole-Parmer® homogenizer
  • Choose from a wide range of accessories for your homogenizer
  • Use with adjustable clamp 04605-08
  • Helps keep samples cold during the grinding process
PRODUCT OPTIONS
£79.63GBP / Each
£95.56GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Use with GeneratorsAdjustable clamp 04605-08
  • DescriptionCryoplate; 1/EA
Key Features
  • Choose from a wide range of accessories for your homogenizer
  • Use with adjustable clamp 04605-08
  • Helps keep samples cold during the grinding process
More About this Item
Optimize your homogenizer with these quality accessories.
Place the cryoplate on top of the vials or microplates loaded in the adjustable clamp. Simply fasten the clamp then begin the grinding process. The cryoplate can also be used with cryo-blocks or insert 04575-82.