Cole-Parmer Spex® 2160 Acid Washed Silica Grinding Beads, 800 to 1,000 µm; 200 g Bottle
Representative image only.
Cole-Parmer Item # WZ-04575-47
Treated to remove fine particles and contaminants
  • Choose from silica or zirconia grinding beads to help remove fine particles and contaminant
  • Suitable for most basic cell lysis applications
  • 800 to 1,000 µm bead size
  • 200 g bottle
PRODUCT OPTIONS
£164.26GBP / Each
£197.11GBP (incl VAT)
SPECS & DESCRIPTION
Specifications & Description
  • Particle Size (micron)800 to 1000
  • MaterialSilica
  • DescriptionAcid Washed Silica Grinding Beads, 800 to 1,000 µm; 200 g Bottle
Key Features
  • Choose from silica or zirconia grinding beads to help remove fine particles and contaminant
  • Suitable for most basic cell lysis applications
  • 800 to 1,000 µm bead size
  • 200 g bottle
More About this Item
Using untreated beads which contain impurities can affect homogenization and biological processes. Acid washing silica and zirconia grinding beads help remove fine particles and contaminant for cleaner samples. Select silica beads for general grinding applications; zirconia beads have greater density and offer higher impact power. General-purpose grinding beads are suitable for most basic cell lysis applications. Choose RNase/DNase-free treated molecular biology-grade grinding beads to inactivate contaminating enzymes. Low-binding grinding beads are coated to reduce nonspecific binding of nucleic acids and proteins.