Speed (rpm)800 to 1200 (1050 for tungsten carbide dish)
Motor Size (hp)1/2
Power (VAC)115
Power (Hz)60
Height (in)54.3
Width (in)26.6
Height (cm)138
Width (cm)67.5
DescriptionShatterBox® Verastile Ring and Puck Mill; 115 VAC, 60 Hz
Warranty3 Year
Includes
ShatterBox, instruction manual, USB memory stick, 10 mm wrench, Torx keys, and power cable.
Key Features
Store up to 10 different grinding programs
Robust IP65 rated industrial interface
Accommodates various grinding dishes
Sound-insulated steel cabinet
Electronically controlled, fully reproducible grinding methods with Ethernet capabilities
3 year warranty
More About this Item
The Cole-Parmer® SM-300 ShatterBox® is a versatile ring and puck mill that can crush and grind many types and sizes of samples with minimal sample loss and increased safety. It features an automated locking dish and guided dish positioning with an automatic elevation of the dish for ease of use and greater flexibility. The versatility of sample containers allows for grinding within smaller dishes without the ring as well as the ability to increase sample sizes and capacity with the ring and puck system.
The ShatterBox swings a dish-shaped grinding container with a puck and ring inside in a tight high-speed circle. The sample is quickly crushed between the wall and floor of the container and the moving puck/ring grinding elements. Grinding times range from two to five minutes, with resultant particle size well below 100 µg/mL, and in some cases 5 µg/mL.
Typical samples include cement, superalloy, granite, glass, sand, slag, ceramics, seeds, grains, pigments, and catalyst supports.
The ShatterBox requires use of a dish with a handle assembly, which is available in hardened steel, tungsten carbide, alumina ceramic, and zirconia. Purchase grinding dish separately.
Effective September 2023, the Spex® SamplePrep 8550 ShatterBox® has now been rebranded Cole-Parmer® under model number SM-300 ShatterBox®. All the same great features and specifications, just with a new and improved look.
Our specialists are here to help you find the best product or part available for your application. Call or Email us and we will make sure you get the right product or part for the job.