This item is now discontinued. Please browse similar items
Spectrum Chromatography 124122 Column Adjustable Plunger 2.5 cm Column ID
Representative image only.
Spectrum Chromatography Mfr # 124122 – Item # WZ-34697-64
Reduce the bed height of your column
  • 3 month warranty
SPECS & DESCRIPTION
Specifications & Description
  • DescriptionColumn Adjustable Plunger 2.5 cm Column ID
  • Warranty3 month
3 Month Warranty
Key Features
  • 3 month warranty
More About this Item
Adjustable Plungers are adaptable bed supports designed to reduce the bed height of a column. Use at either or both ends of the column when you do not want space at the top of the packing. Plungers use the same non-clogging mesh bed support as the columns.
Plungers for aqueous solutions have polypropylene, nylon, PTFE, and Viton® wetted materials. Plungers for organic solutions have only PTFE wetted materials for ultimate chemical resistance.