This item is now discontinued. Please browse similar items
Snyder 5750101N45 Open Top Tank, conical bottom, 330GAL
Representative image only.
Snyder Mfr # 5750101N45 – Item # WZ-06960-70
Open top tank systems provide maximum chemical handling options
  • 3 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • MaterialHDLPE
  • Volume (Gal)330
  • Volume (Liters)1249
  • Height (in)86
  • Height (cm)218.44
  • Outside Diameter (OD) (in)54
  • Graduations (Gal)25
  • Graduations (Liters)95
  • DescriptionOpen Top Tank, conical bottom, 330GAL
  • Warranty3 Years
3 Year Warranty
Key Features
  • 3 year warranty
More About this Item
Available in flat-bottom and total-drain conical bottom, HDLPE tanks are FDA compliant and meet NSF standard 61. Open-top tanks come equipped with a standard lid cover and molded-in gallon/liter graduation indicators. Lids have an all-plastic hinge and incorporate rib design to better support top-fitting installations. Lid assemblies enable access to tank interior without removing entire lid. Conical bottom tanks are designed to withstand weight of 1.9 specific gravity material in tanks. To accommodate floor plumbing and pumps, tank stands have a nominal clearance of 18” from bottom of tank to floor. Tanks do not come with stands, order separately.