This item is now discontinued. Please browse similar items
J. H. Williams Tool Group WSC-250CA Torque Wrench, Split Beam, 45 to 250 Lbs.
Representative image only.
J. H. Williams Tool Group Mfr # WSC-250CA – Item # WZ-25301-27
Rugged design is ideal for any environment
  • Split-beam measuring element provides accurate, reliable readings and eliminates heavy coil spring used in conventional click wrenches
  • Accurate within ±4% of any clockwise setting from 20% of full scale to full scale
  • Sealed head design flexes up to 15°, providing knuckle and obstruction clearance
  • Thumb screw adjustment allows rapid cycling through micrometer-style wrenches
  • Guard prevents accidental setting changes
  • Conversion table to N-m is displayed on the handle
  • Sealed neck keeps dust and grit away from torque mechanism for longer tool life
  • Fewer moving parts reduces friction and wear
  • Ergonomic cushion grip handle provides comfort and control and resists most fluids
  • 1 year warranty
SPECS & DESCRIPTION
Specifications & Description
  • Capacity (lb-ft)250
  • Drive (in)0.5
  • Length (in)21.5
  • DescriptionTorque Wrench, Split Beam, 45 to 250 Lbs.
  • Warranty1 year
1 Year WarrantyConformite Europeenne
Key Features
  • Split-beam measuring element provides accurate, reliable readings and eliminates heavy coil spring used in conventional click wrenches
  • Accurate within ±4% of any clockwise setting from 20% of full scale to full scale
  • Sealed head design flexes up to 15°, providing knuckle and obstruction clearance
  • Thumb screw adjustment allows rapid cycling through micrometer-style wrenches
  • Guard prevents accidental setting changes
  • Conversion table to N-m is displayed on the handle
  • Sealed neck keeps dust and grit away from torque mechanism for longer tool life
  • Fewer moving parts reduces friction and wear
  • Ergonomic cushion grip handle provides comfort and control and resists most fluids
  • 1 year warranty
More About this Item
These unique split-beam torque wrenches provide consistent, accurate readings in a sturdy maintenance-free design. Wrenches allow you to quickly adjust settings by rotating the knob until the required value is displayed in the torque window. The unique sealed ratchet head requires almost no maintenance — allowing you to spend more time working and less time on maintenance. Wrenches are designed for measuring torque in a clockwise direction only (not reversible).